Assay Details
Target Gene Details
Entrez Gene ID: | 66653 |
Gene Name: | BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like |
Gene Aliases: |
2700059M06Rik, 5730512K07Rik, AU018504, AU022602, BRFU, TFIIIB50 |
Location: |
Chr.8:27123355-27128699 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Brf2 | NM_025686.2 | 1 | 146 | NP_079962.1 |
AK012459.1 | 1 | 147 | ||
AK012680.1 | 1 | 147 | BAB28406.1 | |
AK017767.1 | 1 | 149 | ||
AK028228.1 | 1 | 147 | BAC25827.1 | |
AK083122.1 | 1 | 147 | ||
AK151198.1 | 1 | 147 | BAE30195.1 | |
AK152842.1 | 1 | 166 | ||
BC098232.1 | 1 | 127 | AAH98232.1 |
More Information
Set Membership: |
Intragenic Exonic |