Assay Details
Target Gene Details
Entrez Gene ID: | 66423 |
Gene Name: | coordinator of PRMT5, differentiation stimulator |
Gene Aliases: |
1700029I03Rik, 2410022L05Rik, AA409325, AI256813, C85432, Copr5 |
Location: |
Chr.8:13884787-13890327 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Coprs | NM_025556.3 | NP_079832.1 | ||
AK006497.1 | ||||
AK008882.1 | BAB25950.1 | |||
AK010573.1 | BAB27036.1 | |||
BC029192.1 | AAH29192.1 |