Assay Details
Target Gene Details
Entrez Gene ID: | 11945 |
Gene Name: | ATPase, H+/K+ exchanging, beta polypeptide |
Gene Aliases: |
AV080843 |
Location: |
Chr.8:13386209-13396778 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp4b | NM_009724.2 | NP_033854.1 | ||
AK008180.1 | BAB25514.1 | |||
AK008686.1 | ||||
AK009499.1 | BAB26326.1 | |||
AK009670.1 | BAC25265.1 | |||
AK009832.1 | BAB26530.1 | |||
AK009994.1 | BAB26632.1 | |||
AK010003.1 | BAB26635.1 | |||
AK010030.1 | BAB26654.1 | |||
AK010049.1 | 1 | 35 | BAB26665.1 | |
BC120705.1 | ||||
BC125292.1 |
More Information
Set Membership: |
Intragenic Non-exonic |