Assay Details
Target Gene Details
Entrez Gene ID: | 18983 |
Gene Name: | CCR4-NOT transcription complex, subunit 7 |
Gene Aliases: |
AU022737, CAF-1, Caf1, Pop2 |
Location: |
Chr.8:40492538-40515847 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cnot7 | NM_001271542.1 | 3 | 496 | NP_001258471.1 |
NM_001271543.1 | 3 | 567 | NP_001258472.1 | |
NM_011135.5 | 3 | 567 | NP_035265.1 | |
AK007767.1 | 3 | 541 | BAE43206.1 | |
AK009561.1 | 3 | 555 | ||
AK044535.1 | 2 | 1433 | BAC31969.1 | |
AK132063.1 | 3 | 487 | BAE20968.1 | |
AK135669.1 | 3 | 498 | ||
AK137838.1 | 3 | 532 | ||
AK147048.1 | 3 | 534 | BAE27634.1 | |
AK166449.1 | 3 | 531 | BAE38783.1 | |
BC006021.1 | 3 | 511 | AAH06021.1 |
More Information
Set Membership: |
Intragenic Exonic |