Assay Details
Target Gene Details
Entrez Gene ID: | 170776 |
Gene Name: | CD209c antigen |
Gene Aliases: |
SIGNR2, mSIGNR2 |
Location: |
Chr.8:3940221-3955947 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd209c | NM_130903.3 | 4 | 1289 | NP_570973.1 |
AF373410.1 | 4 | 1289 | AAL13236.1 | |
BC120804.1 | 4 | 479 | ||
BC132635.1 | 4 | 479 | ||
BC145088.1 | 4 | 479 |
More Information
Set Membership: |
Intragenic Exonic |