Assay Details
Target Gene Details
Entrez Gene ID: | 68742 |
Gene Name: | transmembrane protein 219 |
Gene Aliases: |
1110032O16Rik, 2900045G02Rik, IGFBP-3R |
Location: |
Chr.7:126886219-126928680 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem219 | NM_026827.1 | 4 | 561 | NP_081103.1 |
NM_028389.1 | 4 | 558 | NP_082665.1 | |
AK004043.1 | 3 | 521 | BAB23139.1 | |
AK013646.1 | 3 | 363 | BAB28936.1 | |
AK152526.1 | 4 | 523 | BAE31285.1 | |
BC023442.1 | 4 | 536 | AAH23442.1 | |
BC046763.1 | 3 | 703 | AAH46763.1 | |
BP769695.1 |