Assay Details
Target Gene Details
Entrez Gene ID: | 22177 |
Gene Name: | TYRO protein tyrosine kinase binding protein |
Gene Aliases: |
DAP12, KARAP, Ly83 |
Location: |
Chr.7:30413788-30417582 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tyrobp | NM_011662.2 | 5 | 485 | NP_035792.1 |
AK154480.1 | 5 | 484 | BAE32616.1 | |
BC056450.1 | 5 | 485 | AAH56450.1 |
More Information
Set Membership: |
Intragenic Exonic |