Assay Details
Target Gene Details
Entrez Gene ID: | 22177 |
Gene Name: | TYRO protein tyrosine kinase binding protein |
Gene Aliases: |
DAP12, KARAP, Ly83 |
Location: |
Chr.7:30413788-30417582 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tyrobp | NM_011662.2 | NP_035792.1 | ||
AF024637.1 | AAD09438.1 | |||
AF077829.1 | AAC95529.1 | |||
AK154480.1 | BAE32616.1 | |||
BC056450.1 | AAH56450.1 |
More Information
Set Membership: |
Intragenic Non-exonic |