Assay Details
Target Gene Details
Entrez Gene ID: | 52377 |
Gene Name: | reticulocalbin 3, EF-hand calcium binding domain |
Gene Aliases: |
6030455P07Rik, D530026G20Rik, D7Ertd671e, RLP49 |
Location: |
Chr.7:45082914-45092257 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rcn3 | NM_026555.2 | 4 | 619 | NP_080831.2 |
AK003918.1 | 3 | 289 | BAB23076.1 | |
AK077943.1 | 4 | 559 | BAC37077.1 | |
AK082762.1 | 4 | 549 | BAC38608.1 | |
AK086705.1 | 4 | 559 | ||
AK132143.1 | 4 | 551 | ||
BC005487.1 | 3 | 456 | AAH05487.1 | |
BC025602.1 | 4 | 570 | AAH25602.1 | |
BC055903.1 | 4 | 566 | AAH55903.1 | |
BC092069.1 | 4 | 517 | AAH92069.1 |
More Information
Set Membership: |
Intragenic Exonic |