Assay Details
Target Gene Details
Entrez Gene ID: | 76560 |
Gene Name: | protease, serine 8 (prostasin) |
Gene Aliases: |
2410039E18Rik, AI313909, C79772, CAP1, fr, mCAP1 |
Location: |
Chr.7:127925717-127930142 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss8 | NM_133351.3 | NP_579929.1 | ||
AB038244.1 | BAB82496.1 | |||
AF188613.1 | AAG17054.1 | |||
AF378085.2 | AAL06319.1 | |||
AK010640.1 | BAC25310.1 | |||
AK078696.1 | BAC37362.1 | |||
AY335911.1 | AAQ91197.1 | |||
BC003851.1 | AAH03851.1 | |||
CB600036.1 |
More Information
Set Membership: |
Intragenic Non-exonic |