Assay Details
Target Gene Details
Entrez Gene ID: | 68845 |
Gene Name: | PIH1 domain containing 1 |
Gene Aliases: |
1110061L23Rik, 4933413A04Rik, Nop17 |
Location: |
Chr.7:45153835-45160065 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pih1d1 | NM_001278207.1 | 9 | 875 | NP_001265136.1 |
NM_001285904.1 | 9 | 867 | NP_001272833.1 | |
NM_029406.4 | 9 | 995 | NP_083682.1 | |
AK004341.1 | 9 | 876 | BAB23269.1 | |
AK016797.1 | 9 | 901 | BAB30435.1 | |
AK089003.1 | 9 | 944 | BAC40694.1 | |
AK146039.1 | 9 | 933 | BAE26851.1 | |
AK152695.1 | 9 | 864 | BAE31425.1 | |
BC068254.1 | 9 | 884 | AAH68254.1 |