Assay Details
Target Gene Details
Entrez Gene ID: | 68845 |
Gene Name: | PIH1 domain containing 1 |
Gene Aliases: |
1110061L23Rik, 4933413A04Rik, Nop17 |
Location: |
Chr.7:45153835-45160065 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pih1d1 | NM_001278207.1 | NP_001265136.1 | ||
NM_001285904.1 | NP_001272833.1 | |||
NM_029406.4 | NP_083682.1 | |||
AK004341.1 | BAB23269.1 | |||
AK016797.1 | BAB30435.1 | |||
AK089003.1 | BAC40694.1 | |||
AK146039.1 | BAE26851.1 | |||
AK152695.1 | BAE31425.1 | |||
AK180877.1 | ||||
BC068254.1 | AAH68254.1 |