Assay Details
Target Gene Details
Entrez Gene ID: | 66985 |
Gene Name: | Ras association (RalGDS/AF-6) domain family (N-terminal) member 7 |
Gene Aliases: |
2400009B11Rik, AW210608 |
Location: |
Chr.7:141215560-141218658 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rassf7 | AK010305.1 |