Assay Details
Target Gene Details
Entrez Gene ID: | 68988 |
Gene Name: | PRP31 pre-mRNA processing factor 31 homolog (yeast) |
Gene Aliases: |
1500019O16Rik, 2810404O06Rik, AW554706, PRP31, RP11 |
Location: |
Chr.7:3629985-3642485 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prpf31 | NM_001159714.1 | NP_001153186.1 | ||
NM_027328.4 | NP_081604.3 | |||
AK005294.1 | BAC25109.1 | |||
AK033190.1 | BAC28192.1 | |||
AK033283.1 | BAC28220.1 | |||
AK044457.1 | BAC31931.1 | |||
AK051260.1 | BAC34578.1 | |||
AY040823.1 | AAK77987.1 | |||
BB839183.1 | ||||
BC018376.1 | AAH18376.1 | |||
BC057877.1 | AAH57877.1 | |||
BC061461.1 | AAH61461.1 | |||
CT010189.1 | CAJ18397.1 |
More Information
Set Membership: |
Intragenic Non-exonic |