Assay Details
Target Gene Details
Entrez Gene ID: | 67773 |
Gene Name: | K(lysine) acetyltransferase 8 |
Gene Aliases: |
2010203C02Rik, 5830450F21Rik, D7Ertd629e, MOF, Myst1 |
Location: |
Chr.7:127912517-127925763 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat8 | NM_026370.1 | NP_080646.1 | ||
AK003264.1 | BAB22680.1 | |||
AK008418.1 | ||||
AK017161.1 | ||||
AK018002.1 | 11 | 1495 | BAC25539.1 | |
AK132000.1 | 11 | 1500 | BAE20930.1 | |
AK146710.1 | BAE27376.1 | |||
AK209737.1 | ||||
BC036284.1 | AAH36284.1 |
Target Gene Details
Entrez Gene ID: | 76560 |
Gene Name: | protease, serine 8 (prostasin) |
Gene Aliases: |
2410039E18Rik, AI313909, C79772, CAP1, fr, mCAP1 |
Location: |
Chr.7:127925717-127930142 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss8 | NM_133351.3 | NP_579929.1 | ||
AB038244.1 | BAB82496.1 | |||
AF188613.1 | AAG17054.1 | |||
AF378085.2 | AAL06319.1 | |||
AY335911.1 | AAQ91197.1 |
More Information
Set Membership: |
Intragenic Non-exonic |