Assay Details
Target Gene Details
Entrez Gene ID: | 212503 |
Gene Name: | polyamine oxidase (exo-N4-amino) |
Gene Aliases: |
2410012F02Rik, AI118225, Pao |
Location: |
Chr.7:140115534-140134368 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Paox | NM_153783.4 | NP_722478.2 | ||
AF226656.2 | AAN40705.2 | |||
AK010469.1 | ||||
AK030664.1 | BAC27070.1 | |||
AK159137.1 | BAE34849.1 | |||
AM049402.1 | CAJ18113.1 | |||
BC033913.1 | AAH33913.1 | |||
BC082783.1 | AAH82783.1 |
More Information
Set Membership: |
Intragenic Non-exonic |