Assay Details
Target Gene Details
Entrez Gene ID: | 114640 |
Gene Name: | parathyroid hormone 2 |
Gene Aliases: |
Tifp39, Tip39 |
Location: |
Chr.7:45176565-45181838 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pth2 | NM_053256.2 | NP_444486.1 | ||
AF399674.1 | AAK84862.1 |