Assay Details
Target Gene Details
Entrez Gene ID: | 16866 |
Gene Name: | luteinizing hormone beta |
Gene Aliases: |
LH, LH-B, LSH-B, LSH-beta |
Location: |
Chr.7:45418166-45421855 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lhb | NM_008497.2 | NP_032523.2 | ||
AV173269.2 | ||||
BB618381.1 | ||||
BM224915.2 | 2 | 462 | ||
Y10418.1 | CAA71445.1 |
More Information
Set Membership: |
Intragenic Non-exonic |