Assay Details
Target Gene Details
Entrez Gene ID: | 269951 |
Gene Name: | isocitrate dehydrogenase 2 (NADP+), mitochondrial |
Gene Aliases: |
E430004F23, IDPm, Idh-2 |
Location: |
Chr.7:80094846-80115387 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Idh2 | NM_173011.2 | NP_766599.2 | ||
AF212319.1 | AAG43538.1 | |||
AK088110.1 | BAC40149.1 | |||
AK145753.1 | BAE26628.1 | |||
AK146564.1 | ||||
AK161640.1 | BAE36505.1 | |||
AK169395.1 | BAE41142.1 | |||
AK205677.1 | ||||
BC060030.1 | AAH60030.1 |
More Information
Set Membership: |
Intragenic Non-exonic |