Assay Details
Target Gene Details
Entrez Gene ID: | 56538 |
Gene Name: | kallikrein related-peptidase 11 |
Gene Aliases: |
2310015I08Rik, Prss20, TLSP |
Location: |
Chr.7:43774617-43779263 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klk11 | NM_001177373.1 | NP_001170844.1 | ||
NM_019974.2 | NP_064358.1 | |||
AB016226.1 | BAA88825.1 | |||
AB016227.1 | BAA36955.1 | |||
AK009360.1 | BAB26241.2 | |||
AK009451.1 | BAB26297.1 | |||
AK009659.1 | ||||
AK009720.1 | BAB26461.2 | |||
AK019102.1 | BAB31548.1 | |||
BC148310.2 |
More Information
Set Membership: |
Intragenic Non-exonic |