Assay Details
Target Gene Details
Entrez Gene ID: | 93747 |
Gene Name: | enoyl Coenzyme A hydratase, short chain, 1, mitochondrial |
Gene Aliases: |
C80529 |
Location: |
Chr.7:140105723-140116423 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Echs1 | NM_053119.2 | NP_444349.1 | ||
AK040391.1 | BAC30583.1 | |||
AK044954.1 | BAC32157.1 | |||
AK086762.1 | BAC39739.1 | |||
AK088018.1 | BAC40099.1 | |||
AK154752.1 | ||||
AK167404.1 | BAE39493.1 | |||
AK210204.1 | ||||
BC002178.1 | AAH02178.1 | |||
BC057971.1 | AAH57971.1 | |||
BC072658.1 | AAH72658.1 |
More Information
Set Membership: |
Intragenic Non-exonic |