Assay Details
Target Gene Details
Entrez Gene ID: | 93747 |
Gene Name: | enoyl Coenzyme A hydratase, short chain, 1, mitochondrial |
Gene Aliases: |
C80529 |
Location: |
Chr.7:140105723-140116423 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Echs1 | NM_053119.2 | NP_444349.1 | ||
AK040391.1 | BAC30583.1 | |||
AK044954.1 | BAC32157.1 | |||
AK086762.1 | BAC39739.1 | |||
AK154752.1 | ||||
AK158306.1 | ||||
BC072658.1 | AAH72658.1 |
Target Gene Details
Entrez Gene ID: | 212503 |
Gene Name: | polyamine oxidase (exo-N4-amino) |
Gene Aliases: |
2410012F02Rik, AI118225, Pao |
Location: |
Chr.7:140115534-140134368 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Paox | BC033913.1 | AAH33913.1 |