Assay Details
Target Gene Details
Entrez Gene ID: | 101502 |
Gene Name: | hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7 |
Gene Aliases: |
AI195443, BB098564 |
Location: |
Chr.7:127800609-127803802 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsd3b7 | NM_001040684.1 | NP_001035774.1 | ||
NM_133943.2 | NP_598704.2 | |||
AK077476.1 | ||||
BB854484.1 | ||||
BY133253.1 |
More Information
Set Membership: |
Intragenic Non-exonic |