Assay Details
Target Gene Details
Entrez Gene ID: | 14356 |
Gene Name: | translocase of inner mitochondrial membrane 10B |
Gene Aliases: |
Fxc1, Tim10b, Tim9b |
Location: |
Chr.7:105640250-105641849 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Timm10b | NM_019502.2 | NP_062375.1 | ||
AF150103.1 | 1 | 65 | AAD40009.1 | |
AK003368.1 | BAB22742.1 | |||
AK003421.1 | ||||
AK010994.1 | 1 | 89 | BAB27313.1 | |
BC009158.1 | AAH09158.1 |
More Information
Set Membership: |
Intragenic Non-exonic |