Assay Details
Target Gene Details
Entrez Gene ID: | 13871 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 2 |
Gene Aliases: |
AA407812, AU020867, AW240756, CXPD, Ercc-2, XPD |
Location: |
Chr.7:19382039-19395692 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ercc2 | NM_007949.4 | 8 | 671 | NP_031975.2 |
AK004652.1 | 4 | 323 | BAB23443.1 | |
AK082323.1 | ||||
AK082761.1 | 8 | 672 | BAC38607.1 | |
AK141692.1 | 8 | 690 | ||
AK145969.1 | 8 | 680 | BAE26794.1 | |
BC034517.1 | 7 | 607 | AAH34517.1 | |
U97572.1 | 7 | 655 | AAB58296.1 |
Target Gene Details
Entrez Gene ID: | 100124438 |
Gene Name: | microRNA 343 |
Gene Aliases: |
Mirn343, mmu-mir-343 |
Location: |
Chr.7:19386643-19386717 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mir343 | NR_030759.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |