Assay Details
Target Gene Details
Entrez Gene ID: | 70394 |
Gene Name: | kaptin |
Gene Aliases: |
2310042D10Rik, 2E4, C030013F01Rik |
Location: |
Chr.7:16119876-16127516 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kptn | NM_133727.2 | NP_598488.2 | ||
AK009746.1 | ||||
AK081226.1 | 2 | 473 | ||
BC018316.1 | AAH18316.1 |