Assay Details
Target Gene Details
Entrez Gene ID: | 56347 |
Gene Name: | eukaryotic translation initiation factor 3, subunit C |
Gene Aliases: |
110kDa, 3230401O13Rik, Eif3s8, NIPIL(A3), NipilA3, Xs, Xsl |
Location: |
Chr.7:126546911-126566366 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif3c | NM_146200.1 | NP_666312.1 | ||
AK088076.1 | ||||
AK152693.1 | ||||
AK160078.1 | BAE35610.1 | |||
AK168160.1 | ||||
AK168174.1 | ||||
AK212354.1 | ||||
BC024855.1 | AAH24855.1 | |||
BC025032.1 | AAH25032.1 | |||
BC080763.1 | AAH80763.1 | |||
U67328.1 | AAB39607.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |