Assay Details
Target Gene Details
Entrez Gene ID: | 67425 |
Gene Name: | EPS8-like 1 |
Gene Aliases: |
2310051G19Rik, 4632407K17Rik, AW060268, DRC3, EPS8R1 |
Location: |
Chr.7:4460631-4480487 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eps8l1 | NM_001290416.1 | NP_001277345.1 | ||
NM_026146.4 | NP_080422.1 | |||
AK009930.1 | ||||
AK014556.1 | ||||
AK019490.2 | BAB31756.2 |
More Information
Set Membership: |
Intragenic Non-exonic |