Assay Details
Target Gene Details
Entrez Gene ID: | 19144 |
Gene Name: | kallikrein related-peptidase 6 |
Gene Aliases: |
AI849898, BSP, Bssp, Klk29, MSP, Prss18, Prss9, neurosin |
Location: |
Chr.7:43824544-43832027 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klk6 | NM_001164696.1 | NP_001158168.1 | ||
NM_001164697.1 | NP_001158169.1 | |||
NM_001164698.1 | NP_001158170.1 | |||
NM_011177.2 | NP_035307.2 | |||
AB008928.1 | BAB55605.1 | |||
AB015206.1 | BAA28895.1 | |||
AK138218.1 | BAE23586.1 | |||
BC031119.1 | AAH31119.1 | |||
EL956218.1 | ||||
Y18723.1 | CAA77269.1 |
More Information
Set Membership: |
Intragenic Non-exonic |