Assay Details
Target Gene Details
Entrez Gene ID: | 14256 |
Gene Name: | FMS-like tyrosine kinase 3 ligand |
Gene Aliases: |
Flt3lg, Ly72L |
Location: |
Chr.7:45131184-45136490 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Flt3l | NM_013520.3 | NP_038548.3 | ||
NR_102725.1 | ||||
AK020105.1 | BAB31997.1 | |||
BC019801.1 | AAH19801.1 | |||
CK622267.1 | ||||
EU274583.1 | ||||
L23636.1 | AAA39436.1 | |||
S76459.1 | AAB33069.1 | |||
U04807.1 | AAA18000.1 |
More Information
Set Membership: |
Intragenic Non-exonic |