Assay Details
Target Gene Details
Entrez Gene ID: | 11717 |
Gene Name: | adenosine monophosphate deaminase 3 |
Gene Aliases: |
- |
Location: |
Chr.7:110767837-110812409 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ampd3 | NM_001276301.1 | NP_001263230.1 | ||
NM_009667.3 | NP_033797.2 | |||
AK133465.1 | BAE21671.1 | |||
BC007183.1 | AAH07183.1 | |||
BC040366.1 | AAH40366.1 | |||
BC056380.1 | AAH56380.1 | |||
D85596.1 | BAA19933.1 |
Target Gene Details
Entrez Gene ID: | 67150 |
Gene Name: | ring finger protein 141 |
Gene Aliases: |
2610110L04Rik, AA792898, AU022812, ZFP36, ZNF230 |
Location: |
Chr.7:110799469-110844452 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnf141 |
More Information
Set Membership: |
Intragenic Non-exonic |