Assay Details
Target Gene Details
Entrez Gene ID: | 11717 |
Gene Name: | adenosine monophosphate deaminase 3 |
Gene Aliases: |
- |
Location: |
Chr.7:110767837-110812409 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ampd3 | NM_001276301.1 | 7 | 1207 | NP_001263230.1 |
NM_009667.3 | 7 | 1326 | NP_033797.2 | |
AK133465.1 | 7 | 1193 | BAE21671.1 | |
BC007183.1 | 2 | 173 | AAH07183.1 | |
BC040366.1 | 7 | 1326 | AAH40366.1 | |
BC056380.1 | 7 | 1168 | AAH56380.1 | |
D85596.1 | 7 | 1205 | BAA19933.1 |
Target Gene Details
Entrez Gene ID: | 67150 |
Gene Name: | ring finger protein 141 |
Gene Aliases: |
2610110L04Rik, AA792898, AU022812, ZFP36, ZNF230 |
Location: |
Chr.7:110799469-110844452 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnf141 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |