Assay Details
Target Gene Details
Entrez Gene ID: | 12337 |
Gene Name: | calpain 5 |
Gene Aliases: |
nCL-3 |
Location: |
Chr.7:98121559-98179298 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Capn5 | NM_007602.4 | 1 | 151 | NP_031628.1 |
AK155179.1 | 1 | 129 | BAE33097.1 | |
AK164293.1 | 1 | 61 | BAE37722.1 | |
AK170034.1 | 1 | 88 | BAE41522.1 | |
BY794575.1 | 1 | 151 | ||
U85020.1 | 1 | 70 | AAD00559.1 | |
Y10656.1 | 1 | 84 | CAA71666.1 |
Target Gene Details
Entrez Gene ID: | 330599 |
Gene Name: | predicted gene, 16938 |
Gene Aliases: |
- |
Location: |
Chr.7:98177194-98184804 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm16938 | NR_045969.1 | |||
NR_045970.1 | ||||
NR_045971.1 | ||||
NR_045972.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |