Assay Details
Target Gene Details
Entrez Gene ID: | 654804 |
Gene Name: | RIKEN cDNA 4732471J01 gene |
Gene Aliases: |
- |
Location: |
Chr.7:25376819-25566417 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4732471J01Rik | NR_015569.3 | |||
AK161422.1 |
Target Gene Details
Entrez Gene ID: | 26365 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 1 |
Gene Aliases: |
Bgp, Bgp1, C-CAM, CD66a, Cc1, Cea-1, Cea-7, Cea1, Cea7, Hv-2, Hv2, MHVR, MHVR1, Mhv-1, bb-1, mCEA1, mmCGM1, mmCGM1a, mmCGM2 |
Location: |
Chr.7:25461702-25477625 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ceacam1 | NM_001039185.1 | NP_001034274.1 | ||
NM_001039186.1 | NP_001034275.1 | |||
NM_001039187.1 | NP_001034276.1 | |||
NM_011926.2 | NP_036056.2 | |||
AB236328.2 | BAE45934.2 | |||
AB236329.1 | BAE45935.1 | |||
AB236330.2 | BAE45936.1 | |||
AB236331.2 | BAE45937.1 | |||
AB236332.2 | BAE45938.1 | |||
M77196.1 | AAA37858.1 | |||
M96934.1 | ||||
M96935.1 | ||||
X15351.1 | CAA33409.1 | |||
X53084.1 | CAA37251.1 | |||
X67278.1 | CAA47695.1 | |||
X67279.1 | CAA47696.1 | |||
X67280.1 | CAA47697.1 | |||
X67281.1 | CAA47698.1 | |||
X67282.1 | CAA47699.1 | |||
X67283.1 | CAA47700.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |