Assay Details
Target Gene Details
Entrez Gene ID: | 260299 |
Gene Name: | cell adhesion molecule 4 |
Gene Aliases: |
Igdf4c, Igsf4c, Tsll2 |
Location: |
Chr.7:24482023-24504533 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cadm4 | NM_153112.3 | NP_694752.1 | ||
AK143204.1 | BAE25303.1 | |||
AY059394.1 | AAL29692.1 |