Assay Details
Target Gene Details
Entrez Gene ID: | 13360 |
Gene Name: | 7-dehydrocholesterol reductase |
Gene Aliases: |
AA409147 |
Location: |
Chr.7:143823167-143848410 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhcr7 | NM_007856.2 | 2 | 431 | NP_031882.1 |
AF057368.1 | AAC40164.1 | |||
AJ307059.1 | 2 | 430 | CAC85215.1 |
More Information
Set Membership: |
Intragenic Exonic |