Assay Details
Target Gene Details
Entrez Gene ID: | 69748 |
Gene Name: | aldehyde dehydrogenase 16 family, member A1 |
Gene Aliases: |
2410004H02Rik, AI452234, mFLJ00209 |
Location: |
Chr.7:45141833-45154538 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aldh16a1 | NM_145954.1 | NP_666066.1 | ||
AK010387.1 | BAB26902.2 | |||
AK030673.1 | BAC27074.1 | |||
AK144995.1 | BAE26175.1 | |||
AK151691.1 | BAE30615.1 | |||
AK153129.1 | BAE31742.1 | |||
AK161198.1 | BAE36234.1 | |||
AK164374.1 | 16 | 2330 | ||
AK170276.1 | BAE41680.1 | |||
AK220200.1 | BAD90125.1 | |||
BC013548.1 | AAH13548.2 | |||
BC030467.1 | AAH30467.1 |
More Information
Set Membership: |
Intragenic Non-exonic |