Assay Details
Target Gene Details
Entrez Gene ID: | 330483 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 16 |
Gene Aliases: |
Gm769 |
Location: |
Chr.7:19852097-19871449 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ceacam16 | NM_001033419.2 | 1 | 135 | NP_001028591.2 |
BC141087.1 |