Assay Details
Target Gene Details
Entrez Gene ID: | 66422 |
Gene Name: | dCTP pyrophosphatase 1 |
Gene Aliases: |
2410015N17Rik, AI854235, RS21-C6 |
Location: |
Chr.7:127256959-127260667 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dctpp1 | NM_023203.1 | NP_075692.1 | ||
AF110764.1 | AAF15970.1 | |||
AK003643.1 | BAB22909.1 | |||
AK010508.1 | BAB26992.1 | |||
AK010606.1 | BAB27056.1 | |||
BC004623.1 | AAH04623.1 |
More Information
Set Membership: |
Intragenic Non-exonic |