Assay Details
Target Gene Details
Entrez Gene ID: | 13401 |
Gene Name: | dystrophia myotonica-containing WD repeat motif |
Gene Aliases: |
DMR-N9, Dm9 |
Location: |
Chr.7:19076089-19082776 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dmwd | NM_010058.2 | NP_034188.2 | ||
AK086899.1 | ||||
BC089027.1 | AAH89027.1 | |||
CF169673.1 | ||||
S60312.1 | AAC60663.1 |