Assay Details
Target Gene Details
Entrez Gene ID: | 66349 |
Gene Name: | ATP5S-like |
Gene Aliases: |
2310004L02Rik, C030044E10Rik |
Location: |
Chr.7:25619414-25625550 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp5sl | NM_001290487.1 | NP_001277416.1 | ||
NM_025504.4 | NP_079780.2 | |||
AK002370.1 | BAB22048.1 | |||
AK009150.1 | BAB26107.1 | |||
AK039669.1 | BAC30413.1 | |||
BC024867.1 | AAH24867.1 |