Assay Details
Target Gene Details
Entrez Gene ID: | 57278 |
Gene Name: | basal cell adhesion molecule |
Gene Aliases: |
1200005K12Rik, B-CAM, Gplu, Lu |
Location: |
Chr.7:19756138-19770532 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcam | NM_020486.2 | 12 | 1592 | NP_065232.1 |
AB035511.1 | 12 | 1553 | BAB16053.1 | |
AF109160.1 | 12 | 1592 | AAF14226.1 | |
AF221507.1 | 12 | 1542 | AAF34657.1 | |
AF346663.1 | 12 | 1530 | AAK83237.1 | |
AK075704.1 | 12 | 1557 | BAC35900.1 | |
AK158439.1 | 12 | 1558 | ||
BC004826.1 | 12 | 1576 | AAH04826.1 |