Assay Details
Target Gene Details
Entrez Gene ID: | 70080 |
Gene Name: | immunoglobulin superfamily, member 23 |
Gene Aliases: |
2210010C17Rik |
Location: |
Chr.7:19937305-19950765 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igsf23 | NM_027308.2 | NP_081584.1 | ||
AK033677.1 | BAC28424.1 | |||
AK088927.1 | BAC40656.1 | |||
BC139455.1 | ||||
BC139458.1 |