Assay Details
Target Gene Details
Entrez Gene ID: | 19713 |
Gene Name: | ret proto-oncogene |
Gene Aliases: |
PTC, RET51, RET9, c-Ret |
Location: |
Chr.6:118151748-118197744 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ret | NM_001080780.1 | 8 | 2207 | NP_001074249.1 |
NM_009050.2 | 8 | 2207 | NP_033076.2 | |
AF209436.1 | 8 | 1596 | AAF21033.1 | |
AK044686.1 | 8 | 1794 | BAC32032.1 | |
AK051633.1 | 8 | 1786 | BAC34699.1 | |
AK084977.1 | 8 | 1857 | ||
AY326397.1 | 8 | 1596 | AAP88379.1 | |
BC059012.1 | 8 | 2181 | AAH59012.1 | |
X67812.1 | 8 | 1596 | CAA48013.1 |
More Information
Set Membership: |
Intragenic Exonic |