Assay Details
Target Gene Details
Entrez Gene ID: | 75430 |
Gene Name: | anaphase prompoting complex C subunit 15 |
Gene Aliases: |
3200002M19Rik, 6330414C15Rik, APC15 |
Location: |
Chr.7:101881300-101901849 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Anapc15 | NM_001291348.1 | NP_001278277.1 | ||
NM_001291349.1 | NP_001278278.1 | |||
NM_001291352.1 | NP_001278281.1 | |||
NM_001291353.1 | NP_001278282.1 | |||
NM_027532.4 | NP_081808.2 | |||
AK014309.1 | BAB29261.2 | |||
AK018169.1 | BAB31105.1 | |||
AK135201.1 | BAE22458.1 | |||
AK139635.1 | BAE24090.1 | |||
BB636259.1 | ||||
BC006736.1 | AAH06736.2 | |||
BC080730.1 | AAH80730.1 | |||
CF166358.1 | ||||
DV659149.1 |