Assay Details
Target Gene Details
Entrez Gene ID: | 66870 |
Gene Name: | serpine1 mRNA binding protein 1 |
Gene Aliases: |
1200009K13Rik, 9330147J08Rik, AL022786, Pairbp1 |
Location: |
Chr.6:67266979-67289302 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serbp1 | NM_001113564.1 | 2 | 499 | NP_001107036.1 |
NM_001113565.1 | 2 | 499 | NP_001107037.1 | |
NM_001113566.1 | 2 | 499 | NP_001107038.1 | |
NM_025814.2 | 2 | 499 | NP_080090.2 | |
AK004678.1 | 2 | 499 | BAB23466.1 | |
AK010851.1 | 2 | 469 | ||
AK010860.1 | 2 | 499 | BAB27229.1 | |
AK011051.1 | 2 | 469 | ||
AK045190.1 | 2 | 463 | BAC32255.1 | |
AK088278.1 | 2 | 468 | BAC40253.1 | |
AK131990.1 | 2 | 468 | BAE20924.1 | |
AK135473.1 | ||||
AK139670.1 | 2 | 468 | ||
AK144764.1 | 2 | 497 | BAE26054.1 | |
AK146418.1 | 2 | 468 | BAE27153.1 | |
AK149505.1 | 2 | 471 | BAE28925.1 | |
AK196615.1 | 2 | 89 | ||
AK201223.1 | 2 | 89 | ||
AK217600.1 | 2 | 89 | ||
BC013665.1 | 2 | 453 | AAH13665.1 | |
BC030502.1 | 2 | 461 | AAH30502.1 |
More Information
Set Membership: |
Intragenic Exonic |