Assay Details
Target Gene Details
Entrez Gene ID: | 19283 |
Gene Name: | protein tyrosine phosphatase, receptor type Z, polypeptide 1 |
Gene Aliases: |
AI850339, DSD-1-PG, PTPbeta, PTPzeta, Ptprz, Ptpz, RPTPz, Rptpbeta |
Location: |
Chr.6:22875458-23052916 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprz1 | NM_001081306.1 | NP_001074775.1 | ||
AJ133130.1 | CAB41976.1 | |||
AK081698.1 | BAC38299.1 | |||
BC151071.1 | ||||
BC157965.1 |
More Information
Set Membership: |
Intragenic Non-exonic |