Assay Details
Target Gene Details
Entrez Gene ID: | 72000 |
Gene Name: | lamin tail domain containing 2 |
Gene Aliases: |
1600016N20Rik |
Location: |
Chr.7:141209990-141215195 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lmntd2 | NM_028050.2 | 12 | 1766 | NP_082326.1 |
AK005476.1 | 3 | 353 | BAB24066.1 | |
BC119093.1 | 12 | 1770 | ||
BC119119.1 | 12 | 1770 | ||
BC144834.1 | 12 | 1800 |