Assay Details
Target Gene Details
Entrez Gene ID: | 72000 |
Gene Name: | lamin tail domain containing 2 |
Gene Aliases: |
1600016N20Rik |
Location: |
Chr.7:141209990-141215195 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lmntd2 | NM_028050.2 | 9 | 1224 | NP_082326.1 |
BC119093.1 | 9 | 1228 | ||
BC119119.1 | 9 | 1228 | ||
BC144834.1 | 9 | 1258 |