Assay Details
Target Gene Details
Entrez Gene ID: | 66508 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 1 |
Gene Aliases: |
2400001E08Rik, Pdro, p18 |
Location: |
Chr.7:101905837-101911903 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lamtor1 | NM_025605.3 | NP_079881.2 | ||
AK003509.1 | ||||
AK009320.1 | BAB26214.1 | |||
AK010247.1 | ||||
AK013389.1 | BAB28825.1 | |||
AK019947.1 | BAB31928.1 | |||
AK144476.1 | BAE25909.1 | |||
AK185971.1 | ||||
BC020142.1 | AAH20142.1 | |||
BC092062.1 | AAH92062.1 | |||
BC096412.1 | AAH96412.1 | |||
BC126973.1 |